Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TGF-beta 2 Rabbit anti-Human, Mouse, Polyclonal, Proteintech

Rabbit Polyclonal Antibody
$518.60
Specifications
Antigen | TGF-beta 2 |
---|---|
Concentration | 0.23 mg/mL |
Applications | Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-173-2648
![]() ![]() |
Proteintech Group Inc
199991AP150UL |
150 μL |
Each of 1 for $518.60
|
|
|||||
Description
This antibody is TGFB2 specific not detecting other TGFBs.
Transforming growth factor type beta 2 (TGFB2) is a member of the TGF-beta family of cytokines, which are ubiquitous, multifunctional, and essential for survival. They play important roles in growth and development, inflammation and repair, and host immunity for many different cell types. Human TGFB2 consists of two disulfide-linked, identical subunits each of which is a single-chain polypeptide of 112 residues.Specifications
TGF-beta 2 | |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
Unconjugated | |
Rabbit | |
Human, Mouse | |
P27090, P61812 | |
21808, 7042 | |
Peptide | |
Primary | |
-20°C | |
TGFB2 |
0.23 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
BSC 1 cell growth inhibitor, Cetermin, G TSF, Polyergin, TGF beta 2, TGF beta2, TGFB2, TGF-beta 2, TGFbeta2 | |
TGFB2 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title