Learn More
TGF-beta 2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Novus Biologicals™
Shop All R&D Systems Products

Description
Specifications
Specifications
| Antigen | TGF-beta 2 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Dilution | Western Blot 0.5 - 4 ug/mL |
| Formulation | PBS, 0.5% BSA, 30% Glycerol with 0.01% Thimerosal |
| Gene Accession No. | P61812 |
| Gene Alias | BSC-1 cell growth inhibitor, Cetermin, Glioblastoma-derived T-cell suppressor factor, G-TSF, MGC116892, Polyergin, TGF-beta2, TGF-beta-2, transforming growth factor beta-2, transforming growth factor, beta 2 |
| Gene Symbols | TGFB2 |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.