Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TGF-beta 3 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB643100
Change view
Click to view available options
Quantity:
100 μg
25 μg
500 μg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB643100 100 μg
MAB643SP 25 μg
MAB643500 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. MAB643100 Supplier R&D Systems Supplier No. MAB643100
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody has been used in 2 publications

TGF-beta 3 Monoclonal specifically detects TGF-beta 3 in Multi-Species samples. It is validated for Western Blot, Immunohistochemistry, ELISA Capture (Matched Antibody Pair).

Specifications

Antigen TGF-beta 3
Applications Western Blot, Immunohistochemistry, ELISA
Classification Monoclonal
Clone 44922
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL, Immunohistochemistry 8-25 ug/mL, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. NP_003230.1
Gene Alias ARVD, ARVD1, FLJ16571, LDS5, RNHF, TGFB3, TGFbeta 3, TGF-beta3, TGF-beta-3, transforming growth factor beta-3, transforming growth factor, beta 3
Gene Symbols TGFB3
Host Species Mouse
Immunogen S. frugiperda insect ovarian cell line Sf 21-derived recombinant human TGF-beta 3 Ala301-Ser412 (Tyr340Phe) Accession # NP_003230.1
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7043
Test Specificity Detects TGF-beta 3 from multiple species in ELISAs and Western blots. In ELISAs, does not cross-react with recombinant human (rh) TGF-beta 1or rhTGF-beta 2.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Multi-species
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Supernatant
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.