Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TGF-beta 3 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. AB244NA
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AB244NA 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. AB244NA Supplier R&D Systems Supplier No. AB244NA
Add to Cart
Add to Cart
This item is not returnable. View return policy

Goat Polyclonal Antibody has been used in 3 publications

TGF-beta 3 Polyclonal specifically detects TGF-beta 3 in Multi-Species samples. It is validated for Neutralization.

Specifications

Antigen TGF-beta 3
Applications Neutralization
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Neutralization 1-3 ug/mL
Gene Accession No. P10600
Gene Alias ARVD, ARVD1, FLJ16571, LDS5, RNHF, TGFB3, TGFbeta 3, TGF-beta3, TGF-beta-3, transforming growth factor beta-3, transforming growth factor, beta 3
Gene Symbols TGFB3
Host Species Goat
Immunogen S. frugiperda insect ovarian cell line Sf 21-derived recombinant chicken TGF-beta 3 and recombinant human TGF-beta 3 Ala301-Ser412 (Tyr340Phe) Accession # P10600
Purification Method Protein A or G purified
Quantity 1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7043
Test Specificity Detects TGF-beta 3 in direct ELISAs. In direct ELISAs, less than 10% cross-reactivity with recombinant amphibian TGF-beta 5 is observed and less than 5% cross-reactivity with recombinant human (rh) TGF-beta 1, rhTGF-beta 2 and rhLAP (TGF-beta 1) is observed.
Reconstitution Reconstitute at 1 mg/mL in sterile PBS.
Target Species Multi-species
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.