Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TGF-beta 3 Biotinylated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. BAF243
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BAF243 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. BAF243 Supplier R&D Systems Supplier No. BAF243
Add to Cart
Add to Cart
This item is not returnable. View return policy

Goat Polyclonal Antibody has been used in 2 publications

TGF-beta 3 Polyclonal specifically detects TGF-beta 3 in Multi-Species samples. It is validated for ELISA Detection (Matched Antibody Pair).

Specifications

Antigen TGF-beta 3
Applications ELISA
Classification Polyclonal
Concentration LYOPH
Conjugate Biotin
Dilution ELISA Detection (Matched Antibody Pair) 0.1-0.4 ug/mL
Gene Alias ARVD, ARVD1, FLJ16571, LDS5, RNHF, TGFB3, TGFbeta 3, TGF-beta3, TGF-beta-3, transforming growth factor beta-3, transforming growth factor, beta 3
Gene Symbols TGFB3
Host Species Goat
Immunogen S. frugiperda insect ovarian cell line Sf 21-derived recombinant chicken TGF-beta 3 and recombinant human TGF-beta 3
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7043
Test Specificity Detects TGF-beta 3 in ELISAs and Western blots. In sandwich immunoassays, less than 0.1% cross-reactivity with recombinant human (rh) TGF-beta 1, rhTGF-beta 1.2, and rhTGF-beta 2 is observed.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Multi-species
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.