Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ TGF beta-3 Monoclonal Antibody (C1)
Mouse Monoclonal Antibody
$221.50 - $522.50
Specifications
Antigen | TGF beta-3 |
---|---|
Clone | C1 |
Concentration | 1 mg/mL |
Content And Storage | -20°C |
Applications | Western Blot |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
PIMA541683
|
Invitrogen™
MA541683 |
200 μL |
Each for $522.50
|
|
|||||
PIMA543704
|
Invitrogen™
MA543704 |
20 μL |
Each for $221.50
|
|
|||||
Description
TGF beta-3 Monoclonal Antibody for Western Blot
Transforming growth factor beta 3 (TGF beta 3) is the third member of the transforming growth factor family of cytokines, which also includes TGF beta 1 and beta 2. These cytokines are secreted in precursor form consisting of a bioactive C-terminal domain attached to an N-terminal domain known as latency associated protein (LAP). Cleavage of LAP results in the mature protein, which functions as a disulfide-linked homodimer. As with all members of the family, TGF beta 3 is highly conserved across species, with mouse and human TGF beta 3 demonstrating 100% sequence homology and cross-species activity.The members of this family can be expressed by most cell types and exert pleiotropic effects, which include the suppression of B- and T-cell effector activity, mediation of tissue healing, and suppression of tumor proliferation. The promotion of CD4+CD25+ T-cell expansion is a newly discovered function of the TGF beta cytokines, and indicates an important role in the protection against autoimmunity.Specifications
TGF beta-3 | |
1 mg/mL | |
Western Blot | |
Unconjugated | |
Mouse | |
RUO | |
PBS with 50% glycerol and 0.05% ProClin 300 | |
P10600 | |
7043 | |
Recombinant protein Transforming Growth Factor Beta 3. The antigen corresponds to amino acid range 24-300 of the target protein. | |
Antibody |
C1 | |
-20°C | |
Monoclonal | |
Liquid | |
IgG2b κ | |
Human | |
TGFB3 | |
ARVD; ARVD1; FLJ16571; H-TGF-b-3; LAP; Latency-associated peptide; LDS5; prepro-transforming growth factor beta-3; RNHF; TGF beta 3; Tgfb3; TGF-B3; Tgfb-3; TGF-beta3; TGF-beta-3; Transforming growth factor; transforming growth factor beta 3; transforming growth factor beta precursor; transforming growth factor beta-3; Transforming growth factor beta-3 proprotein; transforming growth factor, beta 3; transforming growth factor-beta3 | |
TGFB3 | |
Primary | |
Protein A/G |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title