Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TGF-beta Antibody (TGFB/510), Alexa Fluor™ 405, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP308739AF405
Description
TGF-beta Monoclonal antibody specifically detects TGF-beta in Human, Mouse, Rat, Bovine, Canine, Hamster, Monkey samples. It is validated for ELISA, Functional AssaySpecifications
TGF-beta | |
Monoclonal | |
Alexa Fluor 405 | |
CED, DPD1, LAP, TGFB, TGFB1, TGFbeta, transforming growth factor beta 1 | |
Full length recombinant human TGF-beta protein | |
100 μg | |
Cytokine Research, Extracellular Matrix | |
7040 | |
Store at 4°C in the dark. | |
IgG1 κ |
ELISA, Functional Assay | |
TGFB/510 | |
50mM Sodium Borate | |
Mouse | |
Protein A or G purified | |
RUO | |
Primary | |
Human, Mouse, Rat, Bovine, Canine | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction