Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TGF-beta Mouse anti-Human, Mouse, Rat, Bovine, Canine, Hamster, Monkey, Clone: TGFB/510, Novus Biologicals™
SDP

Catalog No. p-7234610 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
20 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB122890 20 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB122890 Supplier Novus Biologicals Supplier No. NBP30770820UG
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

TGF-beta Monoclonal specifically detects TGF-beta in Human, Mouse, Rat, Bovine, Canine, Hamster, Monkey samples. It is validated for ELISA, Functional Assay.

Specifications

Antigen TGF-beta
Applications ELISA, Functional Assay
Classification Monoclonal
Clone TGFB/510
Concentration 0.2 mg/ml
Conjugate Unconjugated
Dilution ELISA 0.1-0.4 ug/ml, Functional Assay
Formulation 10 mM PBS with 0.05% BSA
Gene Alias CED, DPD1, LAP, TGFB, TGFB1, TGFbeta, transforming growth factor beta 1
Host Species Mouse
Immunogen Full length recombinant human TGF-beta protein
Purification Method Protein A or G purified
Quantity 20 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7040
Target Species Human, Mouse, Rat, Bovine, Canine, Hamster, Monkey
Content And Storage Store at 4°C.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.