Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TGF-beta Pan Specific Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. AB100NA
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AB100NA 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. AB100NA Supplier R&D Systems Supplier No. AB100NA
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody has been used in 52 publications

TGF-beta Polyclonal specifically detects TGF-beta in Multi-Species samples. It is validated for Western Blot, Immunohistochemistry, Neutralization.

Specifications

Antigen TGF-beta
Applications Western Blot, Immunohistochemistry, Neutralization
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1 μg/mL, Immunohistochemistry 3-15 μg/mL, Neutralization 5 μg/mL
Gene Alias ARVD1, CED, DPD1, LAP, LDS5, RNHF, TGF beta1, TGFB, TGFB1, TGFb1.2, TGFB2, TGFB3, TGFbeta, TGFbeta 1, TGFbeta 1.2, TGFbeta 2, TGFbeta 3, TGFbeta 5, transforming growth factor beta 1
Gene Symbols TGFB1
Host Species Rabbit
Immunogen Recombinant human TGF-beta 1, porcine platelet-derived TGF-beta 1.2, porcine platelet-derived TGF-beta 2, and recombinant amphibian TGF-beta 5
Purification Method Affinity Purified
Quantity 1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7040
Test Specificity Detects TGF-beta 1, TGF-beta 1.2, TGF-beta 2, TGF-beta 3, and TGF-beta 5 in direct ELISAs and Western blots.
Reconstitution Reconstitute at 1 mg/mL in sterile PBS.
Target Species Multi-species
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.