Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TGF beta Polyclonal Antibody, Abbomax

Catalog No. 501251843
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-125-1843 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-125-1843 Supplier Abbomax Inc Supplier No. 5003944
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Positive control: Breast carcinomas (MCF-7), Placenta, intestine Cellular location: Membrane and cytoplasmic.

TGF beta Polyclonal antibody specifically detects TGF beta in Human, Mouse, Rat samples. It is validated for ELISA, Immunohistochemistry (Paraffin), Western Blot

Specifications

Antigen TGF beta
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with proprietary stabilizer and 0.01% sodium azide
Gene TGFB1
Gene Accession No. P01137, P04202, P17246
Gene Alias CED; DPD1; LAP; Latency-associated peptide; prepro-transforming growth factor beta-1; regulatory protein; tgf beta 1; tgf beta 3; tgf beta1; TGFB; Tgfb1; Tgfb-1; TGFB4; TGFbeta; TGF-beta 1; TGFbeta1; TGF-beta1; TGF-beta-1; TGF-beta4; tissue growth factor-beta4; Transforming growth factor; transforming growth factor beta 1; transforming growth factor beta 4; transforming growth factor beta 4 precursor; transforming growth factor beta-1; Transforming growth factor beta-1 proprotein; transforming growth factor beta-1 proprotein; transforming growth factor beta-1; transforming growth factor, beta 1; transforming growth factor, beta 1 (Camurati-Engelmann disease); transforming growth factor, beta-1; transforming growth factor-beta 1; transforming growth factor-beta-1 precursor
Host Species Rabbit
Immunogen A synthetic peptide derived from N-terminus of human TGF-beta protein
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 21803, 59086, 7040
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.