Learn More
Promega TGF beta R1 Kinase Enzyme System, 1mg
Catalog No. PRV6566 Shop All Promega Corporation Products
Encompass
Change view
Click to view available options
Quantity:
1 mg
Catalog No. Quantity
PRV6566 1 mg
1 options
Catalog No. PRV6566 Supplier Promega Supplier No. V6566
Add to Cart
Add to Cart
Request a Quote
TGF beta R1 is a member of the TGF beta receptor subfamily and is a ser/thr protein kinase that forms a heteromeric complex with type II TGF-beta receptors when bound to TGF-beta, transducing the TGF beta signal from the cell surface to the cytoplasm.
  • Easily Screen and Profile TGFβR1 Kinase Inhibitors
  • Includes kinase, substrate and reaction buffer
  • Use with ADP−Glo™ Assay for bioluminescent detection of kinase activity
Specifications
Quantity 1 mg
Content And Storage <-65°C