Learn More
Promega TGF beta R2 Kinase Enzyme System, 10microg
Catalog No. PRV3931 Shop All Promega Corporation Products
Encompass
Change view
Click to view available options
Quantity:
10 μg
Catalog No. Quantity
PRV3931 10 μg
1 options
Catalog No. PRV3931 Supplier Promega Supplier No. V3931
Recombinant human TGF beta R2 (190-end) was expressed by baculovirus in Sf9 cells using an N-terminal GST tag. TGF beta R2 is a member of the TGF beta receptor subfamily and is a ser/thr protein kinase.
  • Easily Screen and Profile TGFβR2 Kinase Inhibitors
  • Includes kinase, substrate and reaction buffer
  • Use with ADP−Glo™ Assay for bioluminescent detection of kinase activity
Specifications
Quantity 10 μg
Content And Storage <-65°C