Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TGF beta Receptor 1 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501980410
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-0410 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-0410 Supplier Bioss Supplier No. BS0638R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The protein encoded by this gene forms a heteromeric complex with type II TGF-beta receptors when bound to TGF-beta, transducing the TGF-beta signal from the cell surface to the cytoplasm. The encoded protein is a serine/threonine protein kinase. Mutations in this gene have been associated with Loeys-Dietz aortic aneurysm syndrome (LDAS).

Specifications

Antigen TGF beta Receptor 1
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene TGFBR1
Gene Accession No. P36897
Gene Alias AAT5; activin A receptor type II-like kinase, 53kDa; Activin A receptor type II-like protein kinase of 53kD; Activin receptor-like kinase 5; ACVRLK4; ALK5; ALK-5; AU017191; BOS_9013; ESK2; ESS1; LDS1; LDS1A; LDS2A; MSSE; mutant transforming growth factor beta receptor I; ser; Serine/threonine-protein kinase receptor R4; SKR4; TbetaRI; TbetaR-I; TGF-beta receptor type I; TGF-beta receptor type-1; TGF-beta type I receptor; TGFBR1; TGFR1; TGFR-1; Transfor; transforming growth factor beta receptor 1; transforming growth factor beta receptor I; transforming growth factor beta, receptor 1; transforming growth factor, beta receptor 1; transforming growth factor, beta receptor I; transforming growth factor, beta receptor I (activin A receptor type II-like kinase, 53kDa); Transforming growth factor-beta receptor type I
Gene Symbols TGFBR1
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human TGF-beta R1.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7046
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.