Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TGF beta Receptor 2 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501980078
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-0078 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-0078 Supplier Bioss Supplier No. BS0117R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This gene encodes a subunit of mitochondrial ATP synthase. Mitochondrial ATP synthase catalyzes ATP synthesis, utilizing an electrochemical gradient of protons across the inner membrane during oxidative phosphorylation. ATP synthase is composed of two linked multi-subunit complexes: the soluble catalytic core, F1, and the membrane-spanning component, Fo, comprising the proton channel. The catalytic portion of mitochondrial ATP synthase consists of 5 different subunits (alpha, beta, gamma, delta, and epsilon) assembled with a stoichiometry of 3 alpha, 3 beta, and a single representative of the other 3. The proton channel consists of three main subunits (a, b, c). This gene encodes the gamma subunit of the catalytic core. Alternatively spliced transcript variants encoding different isoforms have been identified. This gene also has a pseudogene on chromosome 14.

Specifications

Antigen TGF beta Receptor 2
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene TGFBR2
Gene Accession No. P37173
Gene Alias 1110020H15Rik; AAT3; AU042018; DNIIR; FAA3; LDS1B; LDS2; LDS2B; MFS2; RIIC; RIIDN; TAAD2; TbetaRII; TbetaR-II; TBR-II; TGF-beta 2; TGF-beta receptor II; TGF-beta receptor type II; TGF-beta receptor type IIB; TGF-beta receptor type-2; TGFbeta RII; TGF-beta type II receptor; TGFbeta-RII; Tgfbr2; Tgfbr2T; TGFR2; TGFR-2; transforming growth factor beta receptor 2; transforming growth factor beta receptor II; transforming growth factor beta receptor type II; transforming growth factor beta receptor type IIC; transforming growth factor, beta receptor 2; transforming growth factor, beta receptor II; transforming growth factor, beta receptor II (70/80kDa); transforming growth factor, beta receptor II alpha; transforming growth factor, beta receptor II beta; transforming growth factor, beta receptor II delta; transforming growth factor, beta receptor II epsilon; transforming growth factor, beta receptor II gamma; transforming growth factor, beta receptor IIT; transforming growth factor-b type II receptor; Transforming growth factor-beta receptor type II; transforming growth factor-beta type II receptor
Gene Symbols TGFBR2
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human TGF beta R2.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7048
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.