Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TGF-beta Receptor Rabbit anti-Mouse, Human, Polyclonal, MilliporeSigma™

Catalog No. ABF17IMI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ABF17IMI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ABF17IMI Supplier MilliporeSigma Supplier No. ABF17I
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This rabbit polyclonal Anti-TGF-beta Receptor, type I Immunohistochemistry (Paraffin), and Western Blotting for the detection of TGF-beta receptor type-1.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TGF-beta Receptor
Applications Flow Cytometry, Western Blot, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration Please refer to lot specific datasheet.
Conjugate Unconjugated
Gene Symbols TGFBR1,ALK5,SKR4
Host Species Rabbit
Immunogen KLH-conjugated linear peptide corresponding to 15 amino acids from the C-terminal region of human TGF-beta receptor 1.
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Inflammation and Immunology
Primary or Secondary Primary
Gene ID (Entrez) NP_004603
Test Specificity This polyclonal antibody specifically detects TGF-beta receptor type 1 in multiple species. It targets an epitope of 15 amino acids in the C-terminal region.
Target Species Mouse, Human
Content And Storage Stable for 1 year at 2-8°C from date of receipt.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.