Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TGF-beta RI/ALK-5 Antibody (RM0016-3A11), DyLight 350, Novus Biologicals™

Rat Monoclonal Antibody
Supplier: Novus Biologicals NBP211921UV
Description
TGF-beta RI/ALK-5 Monoclonal specifically detects TGF-beta RI/ALK-5 in Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
TGF-beta RI/ALK-5 | |
Monoclonal | |
DyLight 350 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
TGFBR1 | |
Mouse TGF-beta RI/ALK-5 (extracellular domain) | |
0.1 mL | |
Primary | |
Mouse | |
Purified |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
RM0016-3A11 | |
Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin | |
AAT5, activin A receptor type II-like kinase, 53kD, activin A receptor type II-like kinase, 53kDa, Activin receptor-like kinase 5, ACVRLK4, ALK-5ALK5, EC 2.7.11, EC 2.7.11.30, LDS1A, LDS2A, Serine/threonine-protein kinase receptor R4, SKR4, tbetaR-I, TGFB1R1, TGF-beta receptor type I, TGF-beta receptor type-1, TGF-beta type I receptor, TGFR-1, transforming growth factor beta receptor I, transforming growth factor, beta receptor 1, transforming growth factor, beta receptor I (activin A receptor type II-likekinase, 53kD), Transforming growth factor-beta receptor type I | |
Rat | |
Protein G purified | |
Angiogenesis, Phospho Specific, Protein Kinase | |
7046 | |
Store at 4C in the dark. | |
IgG2 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction