Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TGF-beta RII Antibody (RM0132-6G11) - Azide and BSA Free, Novus Biologicals™
SDP

Rat Monoclonal Antibody

$206.50 - $562.50

Specifications

Antigen TGF-beta RII
Clone RM0132-6G11
Concentration LYOPH
Dilution Western Blot 1:500-1:1000
Applications Western Blot
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB018449
SDP
View Documents
Novus Biologicals
NBP1225080.025MG
0.025 mg
Each for $206.50
Only null left
Add to Cart
 
NBP122508
SDP
View Documents
Novus Biologicals
NBP122508
0.1 mg
Each for $562.50
Only null left
Add to Cart
 
Description

Description

TGF-beta RII Monoclonal specifically detects TGF-beta RII in Mouse samples. It is validated for Western Blot.
Specifications

Specifications

TGF-beta RII
LYOPH
Western Blot
Unconjugated
Rat
Angiogenesis, Cell Cycle and Replication
Q62312
7048
IgG2
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
RM0132-6G11
Western Blot 1:500-1:1000
Monoclonal
Purified
RUO
Mouse
AAT3, EC 2.7.11, EC 2.7.11.30, FAA3, HNPCC6, LDS1B, LDS2B, MFS2, RIIC, TAAD2, tbetaR-II, TGF-beta receptor type II, TGF-beta receptor type IIB, TGF-beta receptor type-2, TGF-beta type II receptor, TGFbeta-RII, TGFR-2, transforming growth factor beta receptor type IIC, transforming growth factor, beta receptor II (70/80kDa), transforming growth factor, beta receptor II (70/80kDa) isoform 1, transforming growth factor, beta receptor II (70/80kDa) isoform 2, transforming growth factor, beta receptor II (70-80kD), Transforming growth factor-beta receptor type II
TGFBR2
Primary
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.