Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TGF-beta RII, Goat anti-Mouse, PE, Polyclonal, R&D Systems™
Goat Polyclonal Antibody
Supplier: R&D Systems FAB532P025
This item is not returnable.
View return policy
Description
Most cell types express three sizes of receptors for TGF-beta. These are designated Type I (53 kDa), Type II (70-85 kDa), and Type III (250-350 kDa). The Type III receptor, a proteoglycan that exists in membrane-bound and soluble forms, binds TGF-beta 1, TGF-beta 2, and TGF-beta 3 but does not appear to be involved in signal transduction. The Type II receptor is a membrane-bound serine/threonine kinase that binds TGF-beta 1 and TGF-beta 3 with high affinity and TGF-beta 2 with a much lower affinity. The Type I receptor is also a membrane-bound serine/threonine kinase that apparently requires the presence of the Type II receptor to bind TGF-beta. Current evidence suggests that signal transduction requires the cytoplasmic domains of both the Type I and Type II receptors.The recombinant soluble TGF-beta Type II receptor is capable of binding TGF-beta 1, TGF-beta 3, and TGF-beta 5 with sufficient affinity to act as an inhibitor of these isoforms at high concentrations. The soluble receptor also binds TGF-beta 2, but with an affinity at least two orders of magnitude lower. Binding of TGF-beta 1, TGF-beta 3, and TGF-beta 5 to the soluble TGF-beta Type II receptor can also be demonstrated by using the soluble receptor as a capture agent on ELISA plates and this observation has been used as the basis for the development of immunoassays for these isoforms of TGF-beta.Specifications
TGF-beta RII | |
Polyclonal | |
Flow Cytometry 10 uL/10^6 cells | |
Q62312 | |
TGFBR2 | |
Mouse myeloma cell line NS0-derived recombinant mouse TGF-beta RII and S. frugiperda insect ovarian cell line Sf 21-derived recombinant mouse TGF-beta RII Ile24-Asp184 Accession # Q62312 | |
25 Tests | |
Primary | |
Detects mouse TGF-beta RII in direct ELISAs and Western blots. In direct ELISAs, approximately 5% cross-reactivity with recombinant human (rh) TGF-beta RII is observed and less than 1% cross-reactivity with recombinant mouse TGF-beta RI and rhTGF-beta RIII is observed. | |
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. |
Flow Cytometry | |
R-PE | |
Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide | |
AAT3, EC 2.7.11, EC 2.7.11.30, FAA3, HNPCC6, LDS1B, LDS2B, MFS2, RIIC, TAAD2, tbetaR-II, TGF-beta receptor type II, TGF-beta receptor type IIB, TGF-beta receptor type-2, TGF-beta type II receptor, TGFbetaRII, TGFbeta-RII, TGFBR2, TGF-bRII, TGFR-2, transforming growth factor beta receptor type IIC, transforming growth factor, beta receptor II (70/80kDa), transforming growth factor, beta receptor II (70/80kDa) isoform 1, transforming growth factor, beta receptor II (70/80kDa) isoform 2, transforming growth factor, beta receptor II (70-80kD), Transforming growth factor-beta receptor type II | |
Goat | |
Affinity Purified | |
RUO | |
7048 | |
Mouse | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction