Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TGF-beta RII Rabbit anti-Human, Clone: 2M8Y2, Novus Biologicals™

Rabbit Monoclonal Antibody
$206.50 - $492.50
Specifications
Antigen | TGF-beta RII |
---|---|
Clone | 2M8Y2 |
Applications | Western Blot |
Classification | Monoclonal |
Conjugate | Unconjugated |
Description
TGF-beta RII Monoclonal antibody specifically detects TGF-beta RII in Human samples. It is validated for Western BlotSpecifications
TGF-beta RII | |
Western Blot | |
Unconjugated | |
Rabbit | |
Angiogenesis, Cell Biology, Cell Cycle and Replication | |
PBS, 0.05% BSA, 50% glycerol, pH7.3 | |
7048 | |
IgG | |
Affinity purified |
2M8Y2 | |
Monoclonal | |
Purified | |
RUO | |
Human | |
AAT3, EC 2.7.11, EC 2.7.11.30, FAA3, HNPCC6, LDS1B, LDS2B, MFS2, RIIC, TAAD2, tbetaR-II, TGF-beta receptor type II, TGF-beta receptor type IIB, TGF-beta receptor type-2, TGF-beta type II receptor, TGFbeta-RII, TGFR-2, transforming growth factor beta receptor type IIC, transforming growth factor, beta receptor II (70/80kDa), transforming growth factor, beta receptor II (70/80kDa) isoform 1, transforming growth factor, beta receptor II (70/80kDa) isoform 2, transforming growth factor, beta receptor II (70-80kD), Transforming growth factor-beta receptor type II | |
Recombinant fusion protein containing a sequence corresponding to amino acids 23-130 of human TGF-beta RII (TGFBR2) (P37173). | |
Primary | |
Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title