Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TGF-beta RIII Antibody (MM0057-5G9), Janelia Fluor™ 646, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP211920JF646
Description
TGF-beta RIII Monoclonal specifically detects TGF-beta RIII in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
TGF-betaRIII | |
Monoclonal | |
Janelia Fluor 646 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
TGFBR3 | |
Human recombinant TGF-beta RIII (extracellular domain) | |
0.1 mL | |
Primary | |
In direct ELISAs, this antibody does not cross-react with recombinant human TGF-beta RI, and TGF-beta RII. | |
Store at 4°C in the dark. | |
IgG1 |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
MM0057-5G9 | |
Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin | |
betaglycan, betaglycan proteoglycan, BGCAN, TGF-beta receptor type 3, TGF-beta receptor type III, TGFR-3, Transforming growth factor beta receptor III, transforming growth factor beta receptor type 3, transforming growth factor, beta receptor III, transforming growth factor, beta receptor III (betaglycan, 300kDa) | |
Mouse | |
Protein G purified | |
Angiogenesis | |
7049 | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction