Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TGFB1 Mouse anti-Human, Clone: B11-4C3, Abnova™

Catalog No. 89035573 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
200 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-035-573 200 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-035-573 Supplier Abnova Corporation Supplier No. MAB14175

Mouse Monoclonal Antibody

Mouse monoclonal antibody raised against full length recombinant human TGFB1.

  • High specificity
  • High reactivity

Specifications

Antigen TGFB1
Applications ELISA, Western Blot
Classification Monoclonal
Clone B11-4C3
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against full length recombinant human TGFB1.
Dilution ELISA (1 ug/mL) Western Blot (1 ug/mL) The optimal working dilution should be determined by the end user.
Formulation In PBS.
Gene Accession No. P01137
Gene Alias CED/DPD1/TGFB/TGFbeta
Gene Symbols TGFB1
Host Species Mouse
Immunogen Recombinant c-terminal polyhistidine fusion protein corresponding to full length human TGFB1.
Purification Method Protein A affinity purification
Quantity 200 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7040
Test Specificity This antibody has successfully been used as the coating antibody in a sandwich ELISA.
Target Species Human
Content And Storage Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG1
Show More Show Less

For Research Use Only (RUO)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.