Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TGFB1 Mouse anti-Human, Clone: E8-4D12, Abnova™
Mouse Monoclonal Antibody
Supplier: Abnova Corporation MAB14176
Description
Mouse monoclonal antibody raised against full length recombinant human TGFB1.
- High specificity
- High reactivity
Specifications
TGFB1 | |
Monoclonal | |
Unconjugated | |
ELISA (1 ug/mL) Western Blot (1 ug/mL) The optimal working dilution should be determined by the end user. | |
P01137 | |
TGFB1 | |
Recombinant c-terminal polyhistidine fusion protein corresponding to full length human TGFB1. | |
200 μg | |
Primary | |
This antibody has successfully been used as the detection antibody in a sandwich ELISA. | |
Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
IgG1 |
ELISA, Western Blot | |
E8-4D12 | |
Mouse monoclonal antibody raised against full length recombinant human TGFB1. | |
In PBS. | |
CED/DPD1/TGFB/TGFbeta | |
Mouse | |
Protein A affinity purification | |
RUO | |
7040 | |
Human | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction