Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Abnova™ Tgfb1 (Rat) ELISA Kit
Click to view available options
Quantity:
1 Kit
Description
Tgfb1 (Rat) ELISA kit is a sandwich enzyme-linked immunosorbent assay for the quantitative measurement of rat Tgfb1 in serum, plasma (heparin,EDTA), cell culture supernatants and urine.
Specifications
Specifications
Description | transforming growth factor, beta 1 |
Cross Reactivity | Rat |
Target Species | Rat |
For Use With (Application) | Quantitative |
Gene ID (Entrez) | 59086 |
Quantity | 1 Kit |
Regulatory Status | RUO |
Storage Requirements | Store at 4°C for 6 months or at -20°C for 12 months. Avoid repeated freeze-thaw cycles. Centrifuge tubes briefly to spin down all components to the bottom before opening. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction