Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TGFB2 Mouse anti-Human, Clone: 220ct16.4.3.1, Abnova™

Catalog No. 89935001 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
400 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-935-001 400 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-935-001 Supplier Abnova Corporation Supplier No. MAB12310
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against human TGFB2.

Specifications

Antigen TGFB2
Applications Immunofluorescence, Immunohistochemistry (PFA fixed), Western Blot
Classification Monoclonal
Clone 220ct16.4.3.1
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against human TGFB2.
Dilution Immunohistochemistry (1:10-1:50) Immunofluorescence (1:10-1:50) Western Blot (1:100-1:1000) The optimal working dilution should be determined by the end user.
Formulation In PBS (0.09% sodium azide)
Gene Alias MGC116892/TGF-beta2
Gene Symbols TGFB2
Host Species Mouse
Immunogen Recombinant protein corresponding to human TGFB2.
Purification Method Protein G purification
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7042
Target Species Human
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.