Learn More
TGFBI Mouse anti-Human, Clone: OTI9A11, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF805390

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
This gene encodes an RGD-containing protein that binds to type I, II and IV collagens. The RGD motif is found in many extracellular matrix proteins modulating cell adhesion and serves as a ligand recognition sequence for several integrins. This protein plays a role in cell-collagen interactions and may be involved in endochondrial bone formation in cartilage. The protein is induced by transforming growth factor-beta and acts to inhibit cell adhesion. Mutations in this gene are associated with multiple types of corneal dystrophy.Specifications
TGFBI | |
Monoclonal | |
Unconjugated | |
TGFBI | |
BIGH3, CDB1, CDG2, CDGG1, CSD, CSD1, CSD2, CSD3, EBMD, LCD1 | |
Mouse | |
Affinity Chromatography | |
RUO | |
7045 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Immunohistochemistry (Paraffin), Western Blot | |
OTI9A11 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
Q15582 | |
TGFBI | |
Human recombit protein fragment corresponding to amino acids 405-683 of human TGFBI produced in E.coli. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.