Learn More
Invitrogen™ TGFBI Polyclonal Antibody

Description
Recommended positive controls: HeLa. Predicted reactivity: Mouse (85%), Rat (85%), Bovine (89%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Specifications
Specifications
| Antigen | TGFBI |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 20% glycerol and 0.01% thimerosal; pH 7 |
| Gene | TGFBI |
| Gene Accession No. | P82198, Q15582 |
| Gene Alias | 68kDa; AI181842; AI747162; beta ig-h3; Beta-ig; beta-ig-h3; betaig-h3; BIGH3; Big-h3; CDB1; CDG2; CDGG1; CSD; CSD1; CSD2; CSD3; EBMD; kerato-epithelin; LCD1; MP70; MP78; RGD-CAP; RGD-containing collagen-associated protein; TGFBI; transforming growth factor beta induced; transforming growth factor beta-induced 68kDa; transforming growth factor induced protein; transforming growth factor, beta induced; transforming growth factor, beta induced, 68 kDa; transforming growth factor, beta-induced, 68kD; transforming growth factor, beta-induced, 68kDa; Transforming growth factor-beta-induced protein ig-h3 |
| Gene Symbols | TGFBI |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.