Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TGFBI Polyclonal Antibody, Invitrogen™

Catalog No. PIPA588096
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA588096 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA588096 Supplier Thermo Scientific Supplier No. PA588096

Rabbit Polyclonal Antibody

Positive Samples: BxPC-3, Mouse heart, Mouse kidney, Rat kidney; Cellular Location: extracellular matrix, extracellular space Immunogen sequence: LPPRERSRLL GDAKELANIL KYHIGDEILV SGGIGALVRL KSLQGDKLEV SLKNNVVSVN KEPVAEPDIM ATNGVVHVIT NVLQPPANRP QERGDELADS A

This gene encodes an RGD-containing protein that binds to type I, II and IV collagens. The RGD motif is found in many extracellular matrix proteins modulating cell adhesion and serves as a ligand recognition sequence for several integrins. This protein plays a role in cell-collagen interactions and may be involved in endochondrial bone formation in cartilage. The protein is induced by transforming growth factor-beta and acts to inhibit cell adhesion. Mutations in this gene are associated with multiple types of corneal dystrophy.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TGFBI
Applications Western Blot
Classification Polyclonal
Concentration 0.66 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.05% ProClin 300, pH 7.3
Gene TGFBI
Gene Accession No. P82198, Q15582
Gene Alias 68kDa, AI181842, AI747162, beta ig-h3, Beta-ig, beta-ig-h3, betaig-h3, BIGH3, Big-h3, CDB1, CDG2, CDGG1, CSD, CSD1, CSD2, CSD3, EBMD, kerato-epithelin, LCD1, MP70, MP78, RGD-CAP, RGD-containing collagen-associated protein, TGFBI, transforming growth factor beta induced, transforming growth factor beta-induced 68kDa, transforming growth factor induced protein, transforming growth factor, beta induced, transforming growth factor, beta induced, 68 kDa, transforming growth factor, beta-induced, 68kD, transforming growth factor, beta-induced, 68kDa, Transforming growth factor-beta-induced protein ig-h3
Gene Symbols TGFBI
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence within amino acids 550-650 of human TGF beta induced (TGFBI) (NP_0003491)
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 21810, 7045
Target Species Human, Mouse, Rat
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.