Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TGFBR2 Polyclonal Antibody, Invitrogen™

Catalog No. PIPA588247
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA588247 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA588247 Supplier Thermo Scientific Supplier No. PA588247

Rabbit Polyclonal Antibody

Immunogen sequence: HNTELLPIEL DTLVGKGRFA EVYKAKLKQN TSEQFETVAV KIFPYEEYAS WKTEKDIFSD INLKHENILQ FLTAEERKTE LGKQYWLITA FHAKGNLQEY LTRHVISWED LRKLGSSLAR GIAHLHSDHT PCGRPKMPIV HRDLKSSNIL VKNDLTCCLC DFGLSLRLDP TLSVDDLANS GQVGTARYMA PEVLESRMNL ENVESFKQTD VYSMALVLWE MTSRCNAVGE VKDYEPPFGS KVREHPCVES MKDNVLRDRG RPEIPSFWLN HQGIQMVCET LTECWDHDPE ARLTAQCVAE RFSELEHLDR LSG Positive Samples: HT-29, A-549, Mouse lung; Cellular Location: Cell membrane, Single-pass type I membrane protein

Transforming Growth Factor, beta receptor 2 (AAT3, FAA3, MFS2, RIIC, LDS1B, LDS2B, TAAD2, TGFR-2, TGFbeta-RII) is a member of the TGF-beta family of receptor Serine/Threonine kinases. It is involved in the regulation of cell proliferation. Mitochondrial ATP synthase catalyzes ATP synthesis, utilizing an electrochemical gradient of protons across the inner membrane during oxidative phosphorylation. ATP synthase is composed of two linked multi-subunit complexes: the soluble catalytic core, F1, and the membrane-spanning component, Fo, comprising the proton channel. The catalytic portion of mitochondrial ATP synthase consists of 5 different subunits (alpha, beta, gamma, delta, and epsilon) assembled with a stoichiometry of 3 alpha, 3 beta, and a single representative of the other 3. The proton channel consists of three main subunits (a, b, c). TGFBR2 encodes the gamma subunit of the catalytic core. Alternatively spliced transcript variants encoding different isoforms have been identified. TGFBR2 also has a pseudogene on chromosome 14.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TGFBR2
Applications Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.44 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal, pH 7.3
Gene TGFBR2
Gene Accession No. P37173, P38438, Q62312
Gene Alias 1110020H15Rik, AAT3, AU042018, DNIIR, FAA3, LDS1B, LDS2, LDS2B, MFS2, RIIC, RIIDN, TAAD2, TbetaRII, TbetaR-II, TBR-II, tgf beta receptor 2, TGF-beta 2, TGF-beta receptor II, TGF-beta receptor type II, TGF-beta receptor type IIB, TGF-beta receptor type-2, TGFbeta RII, TGFbeta type II receptor, TGF-beta type II receptor, TGFbeta-RII, TGFBR2, Tgfbr2T, TGFR2, TGFR-2, transforming growth factor beta receptor 2, transforming growth factor beta receptor II, transforming growth factor beta receptor type II, transforming growth factor beta receptor type IIC, transforming growth factor, beta receptor 2, transforming growth factor, beta receptor II, transforming growth factor, beta receptor II (70/80kDa), transforming growth factor, beta receptor II alpha, transforming growth factor, beta receptor II beta, transforming growth factor, beta receptor II delta, transforming growth factor, beta receptor II epsilon, transforming growth factor, beta receptor II gamma, transforming growth factor, beta receptor IIT, transforming growth factor-b type II receptor, transforming growth factor-beta receptor type II, transforming growth factor-beta type II receptor
Gene Symbols TGFBR2
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 237-549 of human TGF beta Receptor II (TGFBR2) (NP_0032334)
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 21813, 7048, 81810
Target Species Human, Mouse, Rat
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.