Learn More
TGFBR2 Rabbit anti-Human, Polyclonal, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A304413A

Description
The recommended shelf life for this product is 1 year from date of receipt. Based on 100% sequence identity, this antibody is predicted to react with Rat and Chicken
Most mammalian cells express three abundant high affinity receptors, which can bind and be cross-linked to TGF beta: the type I (53 kDa), type II (65 kDa), and type III (100-280 kDa) receptors, based upon the molecular mass of the cross-linked products anSpecifications
TGFBR2 | |
Polyclonal | |
Unconjugated | |
TGFBR2 | |
AAT3, FAA3, LDS1B, LDS2B, MFS2, RIIC, TAAD2, TGFR-2, TGFbeta-RII | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
7048 | |
4° C | |
Liquid |
Immunoprecipitation | |
1 mg/ml | |
phosphate, tris citrate with 0.09% sodium azide; pH 7-8 | |
P37173 | |
TGFBR2 | |
Between 475 and 525 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.