Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TGFBR2 Rabbit anti-Human, Unconjugated, Clone: ABFE-20, Abnova™

Catalog No. 89337176 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-337-176 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-337-176 Supplier Abnova Corporation Supplier No. MAB20776
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody.

TGFBR2 Monoclonal antibody specifically detects TGFBR2 in Human samples. It is validated for WB-Ce.

Specifications

Antigen TGFBR2
Applications Western Blot
Classification Monoclonal
Clone ABFE-20
Conjugate Unconjugated
Description transforming growth factor, beta receptor II (70/80kDa)
Gene TGFBR2
Gene Alias AAT3, FAA3, LDS1B, LDS2B, MFS2, RIIC, TAAD2, TGFR-2, TGFbeta-RII
Host Species Rabbit
Immunogen A synthetic peptide corresponding to human TGFBR2
Quantity 100 μg
Primary or Secondary Primary
Gene ID (Entrez) 7048
Target Species Human
Content And Storage Store at -20°C for one year. After reconstitution, at 4°C for one month. It can also be aliquotted and stored frozen at -20°C for a longer time. Avoid repeated freezing and thawing.
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.