Learn More
TGFBR3 Mouse anti-Human, Clone: OTI6B2, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF809350

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
This locus encodes the transforming growth factor (TGF)-beta type III receptor. The encoded receptor is a membrane proteoglycan that often functions as a co-receptor with other TGF-beta receptor superfamily members. Ectodomain shedding produces soluble TGFBR3, which may inhibit TGFB signaling. Decreased expression of this receptor has been observed in various cancers. Alternatively spliced transcript variants encoding different isoforms have been identified for this gene.Specifications
TGFBR3 | |
Monoclonal | |
Unconjugated | |
TGFBR3 | |
BGCAN, betaglycan | |
Mouse | |
Affinity Chromatography | |
RUO | |
7049 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Western Blot | |
OTI6B2 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
Q03167 | |
TGFBR3 | |
Human recombit protein fragment corresponding to amino acids 531-777 of human TGFBR3 produced in E.coli. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.