Learn More
Invitrogen™ TGFBR3 Polyclonal Antibody
Description
Recommended positive controls: THP-1. Predicted reactivity: Mouse (87%), Rat (88%), Pig (89%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Specifications
Specifications
| Antigen | TGFBR3 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 1.31 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 20% glycerol and 0.025% ProClin 300; pH 7 |
| Gene | TGFBR3 |
| Gene Accession No. | Q03167 |
| Gene Alias | 1110036H20Rik; AU015626; AW215636; betaglycan; betaglycan proteoglycan; BGCAN; OTTHUMP00000011787; TBRIII; tgf beta receptor 3; TGF-beta receptor type 3; TGF-beta receptor type III; Tgfbr3; TGFR-3; transforming growth factor beta receptor 3; transforming growth factor beta receptor III; Transforming growth factor beta receptor type 3; transforming growth factor, beta receptor 3; transforming growth factor, beta receptor III |
| Gene Symbols | TGFBR3 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.