Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TGFBR3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5116035
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5116035 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5116035 Supplier Invitrogen™ Supplier No. PA5116035
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total TGF beta Receptor III.

This locus encodes the transforming growth factor (TGF)-beta type III receptor. The encoded receptor is a membrane proteoglycan that often functions as a co-receptor with other TGF-beta receptor superfamily members. Ectodomain shedding produces soluble TGFBR3, which may inhibit TGFB signaling. Decreased expression of this receptor has been observed in various cancers. Alternatively spliced transcript variants encoding different isoforms have been identified for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TGFBR3
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide
Gene TGFBR3
Gene Accession No. O88393, P26342, Q03167
Gene Alias 1110036H20Rik; AU015626; AW215636; betaglycan; betaglycan proteoglycan; BGCAN; OTTHUMP00000011787; TBRIII; tgf beta receptor 3; TGF-beta receptor type 3; TGF-beta receptor type III; Tgfbr3; TGFR-3; transforming growth factor beta receptor 3; transforming growth factor beta receptor III; Transforming growth factor beta receptor type 3; transforming growth factor, beta receptor 3; transforming growth factor, beta receptor III
Gene Symbols TGFBR3
Host Species Rabbit
Immunogen A synthesized peptide derived from human TGFBR3(Accession Q03167), corresponding to amino acid residues R818-A851.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 21814, 29610, 7049
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.