Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ TGFBRAP1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA558374
Description
Immunogen sequence: RQQLFHTLLA IYLHAGPTAH ELAVAAVDLL NRHATEFDAA QVLQMLPDTW SVQLLCPFLM GAMRDSIHAR RTMQVALGLA RSENLIYTYD KMKLKG Highest antigen sequence identity to the following orthologs: Mouse - 84%, Rat - 85%.
Plays a role in the TGF-beta/activin signaling pathway. It associates with inactive heteromeric TGF-beta and activin receptor complexes, mainly through the type II receptor, and is released upon activation of signaling. May recruit SMAD4 to the vicinity of the receptor complex and facilitate its interaction with receptor-regulated Smads, such as SMAD2.
Specifications
TGFBRAP1 | |
Polyclonal | |
Unconjugated | |
TGFBRAP1 | |
TGF-beta receptor-associated protein 1; TGFBRAP1; transforming growth factor beta receptor associated protein 1; transforming growth factor-beta receptor-associated protein 1; TRAP1; TRAP-1; VPS3; VPS3 CORVET complex subunit | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
9392 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q8WUH2 | |
TGFBRAP1 | |
Recombinant protein corresponding to Human TGFBRAP1. Recombinant protein control fragment (Product #RP-96084). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction