Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TGFBRAP1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA565189
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA565189 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA565189 Supplier Invitrogen™ Supplier No. PA565189
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: SAAWLEKHKKY FALGLLYHYN NQDAAAVQLW VNIVNGDVQD STRSDLYEYI VDFLTYCLDE ELVWAYADWV LQKSEEVGVQ VFTKRPLDEQ QKNSFNPDD Highest antigen sequence identity to the following orthologs - mouse 88%, rat 90%.

Plays a role in the TGF-beta/activin signaling pathway. It associates with inactive heteromeric TGF-beta and activin receptor complexes, mainly through the type II receptor, and is released upon activation of signaling. May recruit SMAD4 to the vicinity of the receptor complex and facilitate its interaction with receptor-regulated Smads, such as SMAD2.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TGFBRAP1
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene TGFBRAP1
Gene Accession No. Q8WUH2
Gene Alias TGF-beta receptor-associated protein 1; TGFBRAP1; transforming growth factor beta receptor associated protein 1; transforming growth factor-beta receptor-associated protein 1; TRAP1; TRAP-1; VPS3; VPS3 CORVET complex subunit
Gene Symbols TGFBRAP1
Host Species Rabbit
Immunogen Recombinant Human TGFBRAP1. Recombinant protein control fragment (Product #RP-104676).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9392
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.