Learn More
TGIF1 Mouse anti-Human, Clone: OTI2E7, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF802353

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
TGIF is a member of the three-amino acid loop extension (TALE) superclass of atypical homeodomains. TALE homeobox proteins are highly conserved transcription regulators. This particular homeodomain binds to a previously characterized retinoid X receptor responsive element from the cellular retinol-binding protein II promoter. In addition to its role in inhibiting 9-cis-retinoic acid-dependent RXR alpha transcription activation of the retinoic acid responsive element, the protein is an active transcriptional co-repressor of SMAD2 and may participate in the transmission of nuclear signals during development and in the adult. Mutations in this gene are associated with holoprosencephaly type 4, which is a structural anomaly of the brain.Specifications
TGIF1 | |
Monoclonal | |
Unconjugated | |
TGIF1 | |
HPE4, TGIF | |
Mouse | |
Affinity Chromatography | |
RUO | |
7050 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Western Blot | |
OTI2E7 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
Q15583 | |
TGIF1 | |
Full length human recombit protein of human TGIF1 produced in E.coli. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.