Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ TGIF2 Monoclonal Antibody (4B10)
Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA522442
Description
Peptide Sequence: LAVSVPAPTN VLSLSVCSMP LHSGQGEKPA APFPRGELES PKPLVTPGST LTLLTRAEAG SP.
The protein encoded by this gene is a DNA-binding homeobox protein and a transcriptional repressor, which appears to repress transcription by recruiting histone deacetylases to TGF beta-responsive genes. This gene is amplified and over-expressed in some ovarian cancers. Alternative splicing results in multiple transcript variants. A related pseudogene has been identified on chromosome 1. Read-through transcription also exists between this gene and the neighboring downstream C20orf24 gene.
Specifications
TGIF2 | |
Monoclonal | |
1 mg/mL | |
PBS with no preservative; pH 7.4 | |
Q9GZN2 | |
TGIF2 | |
TGIF2 (NP_068581.1, 114 a.a. ~ 175 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG2a κ |
ELISA, Western Blot, Immunocytochemistry | |
4B10 | |
Unconjugated | |
TGIF2 | |
4921501K24; 5730599O09Rik; 5'-TG-3' interacting factor 2; 5'-TG-3'-interacting factor 2; C80753; C81206; Homeobox protein TGIF2; OTTHUMP00000030849; RGD1560115; RGD1564927; TALE family homeobox; TGF(beta)-induced transcription factor 2; TGFB induced factor homeobox 2; TGF-beta-induced transcription factor 2; TGFB-induced factor 2; TGFB-induced factor 2 (TALE family homeobox); TGFB-induced factor homeobox 2; TGFB-induced factor homeobox 2-like 1; TGIF2; Tgif2l1; transcription growth factor-beta-induced factor 2 | |
Mouse | |
Affinity chromatography | |
RUO | |
60436 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction