Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ TGIF2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA521900
Description
Recommended positive controls: A431, H1299, HeLa. Predicted reactivity: Rat (100%), Rhesus Monkey (100%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
The protein encoded by this gene is a DNA-binding homeobox protein and a transcriptional repressor, which appears to repress transcription by recruiting histone deacetylases to TGF beta-responsive genes. This gene is amplified and over-expressed in some ovarian cancers. Alternative splicing results in multiple transcript variants. A related pseudogene has been identified on chromosome 1. Read-through transcription also exists between this gene and the neighboring downstream C20orf24 gene.
Specifications
| TGIF2 | |
| Polyclonal | |
| Unconjugated | |
| TGIF2 | |
| 4921501K24; 5730599O09Rik; 5'-TG-3' interacting factor 2; 5'-TG-3'-interacting factor 2; C80753; C81206; Homeobox protein TGIF2; OTTHUMP00000030849; RGD1560115; RGD1564927; TALE family homeobox; TGF(beta)-induced transcription factor 2; TGFB induced factor homeobox 2; TGF-beta-induced transcription factor 2; TGFB-induced factor 2; TGFB-induced factor 2 (TALE family homeobox); TGFB-induced factor homeobox 2; TGFB-induced factor homeobox 2-like 1; TGIF2; Tgif2l1; transcription growth factor-beta-induced factor 2 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 60436 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 0.59 mg/mL | |
| 0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7 | |
| Q9GZN2 | |
| TGIF2 | |
| Carrier-protein conjugated synthetic peptide encompassing a sequence within the C-terminus region of human TGF beta induced factor 2. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction