Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TGIF2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5111379
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5111379 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5111379 Supplier Invitrogen™ Supplier No. PA5111379
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: SVLAVSVPAP TNVLSLSVCS MPLHSGQGEK PAAPFPRGEL ESPKPLVTPG STLTLLTRAE AGSPTGGLF Highest antigen sequence indentity to the following orthologs: Mouse - 91%, Rat - 91%.

The protein encoded by this gene is a DNA-binding homeobox protein and a transcriptional repressor, which appears to repress transcription by recruiting histone deacetylases to TGF beta-responsive genes. This gene is amplified and over-expressed in some ovarian cancers. Alternative splicing results in multiple transcript variants. A related pseudogene has been identified on chromosome 1. Read-through transcription also exists between this gene and the neighboring downstream C20orf24 gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TGIF2
Applications ChIP Assay, Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene TGIF2
Gene Accession No. Q9GZN2
Gene Alias 4921501K24; 5730599O09Rik; 5'-TG-3' interacting factor 2; 5'-TG-3'-interacting factor 2; C80753; C81206; Homeobox protein TGIF2; OTTHUMP00000030849; RGD1560115; RGD1564927; TALE family homeobox; TGF(beta)-induced transcription factor 2; TGFB induced factor homeobox 2; TGF-beta-induced transcription factor 2; TGFB-induced factor 2; TGFB-induced factor 2 (TALE family homeobox); TGFB-induced factor homeobox 2; TGFB-induced factor homeobox 2-like 1; TGIF2; Tgif2l1; transcription growth factor-beta-induced factor 2
Gene Symbols TGIF2
Host Species Rabbit
Immunogen Recombinant Protein corresonding to Human TGIF2. Recombinant protein control fragment (Product #RP-106494).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 60436
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.