Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TGIF2LX Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA556971
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA556971 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA556971 Supplier Invitrogen™ Supplier No. PA556971
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: PDNVQSLPLW PLPKGQMSRE KQPDPESAPS QKLTGIAQPK KKVKVSVTSP SSPELVSPEE HADFSSFLLL VDAAVQRAAE LELEKKQEPN Highest antigen sequence identity to the following orthologs: Mouse - 48%, Rat - 44%.

TGIF2LX is a member of the TALE/TGIF homeobox family of transcription factors. Testis-specific expression suggests that this gene may play a role in spermatogenesis. This gene encodes a member of the TALE/TGIF homeobox family of transcription factors. Testis-specific expression suggests that this gene may play a role in spermatogenesis. A homolog of this gene lies within the male specific region of chromosome Y, in a block of sequence that is thought to be the result of a large X-to-Y transposition.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TGIF2LX
Applications ChIP Assay, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene TGIF2LX
Gene Accession No. Q8IUE1
Gene Alias homeobox protein TGIF2LX; MGC34726; OTTMUSG00000018465; testis expressed homeobox Tex1; Tex1; TGF beta-induced transcription factor 2-like protein; TGFB induced factor homeobox 2 like X-linked; TGFB induced factor homeobox 2 like, X-linked; TGF-beta-induced transcription factor 2-like protein; TGFB-induced factor 2-like protein, X-linked; TGFB-induced factor 2-like, X-linked; TGFB-induced factor homeobox 2-like, X-linked; TGFB-induced factor homeobox 2-like, X-linked 1; TGIF homeobox 1; TGIF2LX; Tgif2lx1; TGIF-like on the X; TGIFLX; Tgifx1
Gene Symbols TGIF2LX
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human TGIF2LX. Recombinant protein control fragment (Product #RP-95452).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 90316
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.