Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TGIF2LY Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA575716
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA575716 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA575716 Supplier Invitrogen™ Supplier No. PA575716
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The antibody was affinity-purified from rabbit antiserum by affinity-chromatography using epitope-specific immunogen and the purity is > 95% (by SDS-PAGE).

TGIF2LY is a member of the TALE/TGIF homeobox family of transcription factors. This gene lies within the male specific region of chromosome Y, in a block of sequence that is thought to be the result of a large X-to-Y transposition.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TGIF2LY
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.2
Gene TGIF2LY
Gene Accession No. Q8IUE0
Gene Alias Homeobox protein TGIF2LY; TGFB induced factor homeobox 2 like, Y-linked; TGF-beta-induced transcription factor 2-like protein; TGFB-induced factor 2-like protein, Y-linked; TGFB-induced factor 2-like, Y-linked; TGFB-induced factor homeobox 2-like, Y-linked; TGIF2LY; TGIF-like on the Y; TGIFLY
Gene Symbols TGIF2LY
Host Species Rabbit
Immunogen Synthetic peptide corresponding to residues in Human TGIF2LX.
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 90655
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.