Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TGIF2LY Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA541207
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA541207 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA541207 Supplier Thermo Scientific Supplier No. PA541207
Add to Cart
Add to Cart

TGIF2LY Polyclonal Antibody

TGIF2LY Polyclonal Antibody for Western Blot

TRUSTED_SUSTAINABILITY

Specifications

Antigen TGIF2LY
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene TGIF2LY
Gene Accession No. Q8IUE0
Gene Alias Homeobox protein TGIF2LY; TGFB induced factor homeobox 2 like, Y-linked; TGF-beta-induced transcription factor 2-like protein; TGFB-induced factor 2-like protein, Y-linked; TGFB-induced factor 2-like, Y-linked; TGFB-induced factor homeobox 2-like, Y-linked; TGIF2LY; TGIF-like on the Y; TGIFLY
Gene Symbols TGIF2LY
Host Species Rabbit
Immunogen Synthetic peptide directed towards the middle region of human TGIF2LY.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 90655
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.