Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TGIF2LX Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA568895
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA568895 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA568895 Supplier Invitrogen™ Supplier No. PA568895
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This target displays homology in the following species: Mouse: 100%; Rat: 86%.

TGIF2LX is a member of the TALE/TGIF homeobox family of transcription factors. Testis-specific expression suggests that this gene may play a role in spermatogenesis. This gene encodes a member of the TALE/TGIF homeobox family of transcription factors. Testis-specific expression suggests that this gene may play a role in spermatogenesis. A homolog of this gene lies within the male specific region of chromosome Y, in a block of sequence that is thought to be the result of a large X-to-Y transposition.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TGIF2LX
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene Tgif2lx1
Gene Accession No. 0
Gene Alias homeobox protein TGIF2LX; MGC34726; OTTMUSG00000018465; testis expressed homeobox Tex1; Tex1; TGF beta-induced transcription factor 2-like protein; TGFB induced factor homeobox 2 like X-linked; TGFB induced factor homeobox 2 like, X-linked; TGF-beta-induced transcription factor 2-like protein; TGFB-induced factor 2-like protein, X-linked; TGFB-induced factor 2-like, X-linked; TGFB-induced factor homeobox 2-like, X-linked; TGFB-induced factor homeobox 2-like, X-linked 1; TGIF homeobox 1; TGIF2LX; Tgif2lx1; TGIF-like on the X; TGIFLX; Tgifx1
Gene Symbols Tgif2lx1
Host Species Rabbit
Immunogen synthetic peptide corresponding to a region of mouse Tgifx1.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 245583
Target Species Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.