Learn More
Invitrogen™ TGM1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5120211
Description
Positive test controls include: Mouse liver, Rat spleen. The target is usually found in the following locations: Lipid-anchor, Membrane. Immunogen sequence: AGDGTIREGM LVVNGVDLLS SRSDQNRREH HTDEYEYDEL IVRRGQPFHM LLLLSRTYES SDRITLELLI GNNPEVGKGT HVIIPVGKGG SGGWKAQVVK ASGQNLNLRV HTSPNAIIGK FQFTVRTQSD AGEFQLPFDP RNEIYILFNP WCPEDIVYVD HEDWRQEYVL NESGRIYYGT E.
Protein kinase C (PKC) is a family of serine- and threonine-specific protein kinases that can be activated by calcium and second messenger diacylglycerol. PKC family members phosphorylate a wide variety of protein targets and are known to be involved in diverse cellular signaling pathways. PKC family members also serve as major receptors for phorbol esters, a class of tumor promoters. Each member of the PKC family has a specific expression profile and is believed to play a distinct role in cells.
Specifications
TGM1 | |
Polyclonal | |
Unconjugated | |
TGM1 | |
2310004J08Rik; ARCI1; Epidermal TGase; ICR2; K polypeptide; K polypeptide epidermal type I, protein-glutamine-gamma-glutamyltransferase; KTG; LI; LI1; LOW QUALITY PROTEIN: protein-glutamine gamma-glutamyltransferase K; Protein-glutamine gamma-glutamyltransferase K; protein-glutamine-gamma-glutamyltransferase; TG K; TG(K); TGASE; TGase 1; TGase K; Tgase1; TGase-1; TGK; TGM1; tranglutaminase 1; transglutaminase 1; transglutaminase 1 (K polypeptide epidermal type I, protein-glutamine-gamma-glutamyltransferase); transglutaminase 1 isoform; transglutaminase 1, K polypeptide; transglutaminase K; transglutaminase type 1; transglutaminase type I; transglutaminase, keratinocyte; transglutaminase-1 | |
Rabbit | |
Affinity chromatography | |
RUO | |
21816, 60335, 7051 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Western Blot | |
1.56 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P22735, P23606, Q9JLF6 | |
TGM1 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 100-280 of human TGM1 (NP_000350.1). | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.