Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TGM2 Mouse anti-Human, Clone: OTI1A3, lyophilized, TrueMAB™
SDP

Catalog No. 501669327
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-166-9327 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-166-9327 Supplier OriGene Supplier No. CF809188
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO

Transglutaminase II catalyzes calcium-dependent post-translational modification of proteins by formation of an isopeptide bond within or between polypeptide chains. It is also known as TGC, tTG, type II-, Gh, cytosolic-, liver-, endothelial-, erythrocyte-, cellular-transglutaminase. Different tissues and cell types express varying amounts of tissue transglutaminase with a markedly hugh expression in rheumatoid lesions. It is implicated in programmed cell death, signal transduction, drug-resistance, cell growth, endocytosis, insulin secretion, cell adhesion, cataract formation, and wound healing.

Specifications

Antigen TGM2
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone OTI1A3
Conjugate Unconjugated
Formulation PBS with 8% trehalose and no preservative; pH 7.3
Gene TGM2
Gene Accession No. P21980
Gene Alias TG(C), TGC
Gene Symbols TGM2
Host Species Mouse
Immunogen Full length human recombit protein of human TGM2 produced in HEK293T cell.
Purification Method Affinity Chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7052
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Lyophilized
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.