Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TGM2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5119473
Change view
Click to view available options
Quantity:
20 μL
200 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5119473 200 μL
PIPA5121549 20 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. PIPA5119473 Supplier Invitrogen™ Supplier No. PA5119473
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

TGM2 Polyclonal Antibody for Western Blot, ICC/IF, IHC (P)

Transglutaminase (TGase) catalyses the crosslink of proteins by forming e-(g-glutamyl) lysine isopeptide bonds and requires the binding of Ca2+ for its activity. In mammals, eight distinct TGase isoenzymes have been identified. Tissue transglutaminase (tTGase), also known as TGase 2, has four distinct domains: N-terminal b-sandwich, catalytic core and two C-terminal b-barrel domains. tTGase may have a role in cell death, cell proliferation, cell differentiation, and receptor-mediated endocytosis. In the Alzheimer's disease brain, the elevated tTGase activity is manifested by polymerization of a number of proteins, including Ab peptide, b-amyloid precursor protein and the tau protein, with formation of neurofibrillary tangles.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TGM2
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4
Gene TGM2
Gene Accession No. P21980, P21981
Gene Alias C polypeptide; C polypeptide, protein-glutamine-gamma-glutamyltransferase; G alpha h; G[a]h; G-ALPHA-h; GNAH; Protein-glutamine gamma-glutamyltransferase 2; protein-glutamine-gamma-glutamyltransferase; RP5-1054A22.2; TG C; TG(C); TG2; TGase C; TGase H; TGase2; TGase-2; TGase-H; TgaseII; TGC; TGM2; Tissue transglutaminase; Tissue type transglutaminase; tissue-type transglutaminase; transglutaminase; transglutaminase 2; transglutaminase 2 (C polypeptide, protein-glutamine-gamma-glutamyltransferase); Transglutaminase 2 C polypeptide; transglutaminase 2, C polypeptide; transglutaminase C; Transglutaminase H; transglutaminase-2; tTG; tTGas
Gene Symbols TGM2
Host Species Rabbit
Immunogen Recombinant protein Transglutaminase 2. The antigen corresponds to amino acid range 1-687 of the target protein.
Purification Method Antigen affinity chromatography, Protein A
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 21817, 56083, 7052
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.