Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TGM2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody
Supplier: Proteintech Group Inc 151001AP150UL

Description
Transglutaminase II catalyzes calcium-dependent post-translational modification of proteins by formation of an isopeptide bond within or between polypeptide chains. It is also known as TGC, tTG, type II-, Gh, cytosolic-, liver-, endothelial-, erythrocyte-, cellular-transglutaminase. Different tissues and cell types express varying amounts of tissue transglutaminase with a markedly hugh expression in rheumatoid lesions. It is implicated in programmed cell death, signal transduction, drug-resistance, cell growth, endocytosis, insulin secretion, cell adhesion, cataract formation, and wound healing.Specifications
TGM2 | |
Polyclonal | |
Unconjugated | |
TGM2 | |
G ALPHA h, GNAH, TG(C), TG2, TGase 2, TGase C, TGase H, TGC, TGM2, Tissue transglutaminase, Transglutaminase 2, Transglutaminase C, Transglutaminase H | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
21817, 56083, 7052 | |
-20°C | |
Liquid |
Immunoprecipitation, Western Blot | |
0.18 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P21980, P21981 | |
TGM2 | |
TGM2 Fusion Protein Ag7439 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction