Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TGM3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA537896
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA537896 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA537896 Supplier Invitrogen™ Supplier No. PA537896
Add to Cart
Add to Cart

Goat Polyclonal Antibody

This antibody is tested in Peptide ELISA: antibody detection limit dilution 32,000.

Transglutaminases are enzymes that catalyze the crosslinking of proteins by epsilon-gamma glutamyl lysine isopeptide bonds. While the primary structure of transglutaminases is not conserved, they all have the same amino acid sequence at their active sites and their activity is calcium-dependent. The protein encoded by this gene consists of two polypeptide chains activated from a single precursor protein by proteolysis. The encoded protein is involved the later stages of cell envelope formation in the epidermis and hair follicle.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TGM3
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3
Gene TGM3
Gene Accession No. Q08188
Gene Alias AI893889; E polypeptide; E polypeptide, protein-glutamine-gamma-glutamyltransferase; MGC126249; MGC126250; protein-glutamine gamma-glutamyltransferase E; Protein-glutamine gamma-glutamyltransferase E 27 kDa non-catalytic chain; Protein-glutamine gamma-glutamyltransferase E 50 kDa catalytic chain; protein-glutamine-gamma-glutamyltransferase; RGD1561831; TG E; TG(E); TGase E; TGase TGase-3; Tgase3; TGase-3; TGE; TGM3; transglutaminase 3; transglutaminase 3 (E polypeptide, protein-glutamine-gamma-glutamyltransferase); transglutaminase 3, E polypeptide; transglutaminase E; Transglutaminase-3
Gene Symbols TGM3
Host Species Goat
Immunogen Peptide with sequence C-TLEVLNEARVRKP, from the internal region of TGM3 (aa598-610).
Purification Method Ammonium sulfate precipitation
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7053
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.