Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ TGM5 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA543311
Description
Peptide sequence: VLKPQHQASI ILETVPFKSG QRQIQANMRS NKFKDIKGYR NVYVDFAL Sequence homology: Cow: 93%; Dog: 100%; Guinea Pig: 100%; Horse: 93%; Human: 100%; Mouse: 86%; Rabbit: 85%; Rat: 93%.
This gene encodes a member of the transglutaminase family. The encoded protein catalyzes formation of protein cross-links between glutamine and lysine residues, often resulting in stabilization of protein assemblies. This reaction is calcium dependent. Mutations in this gene have been associated with acral peeling skin syndrome.
Specifications
TGM5 | |
Polyclonal | |
Unconjugated | |
Tgm5 | |
2310007C07Rik; MGC141907; protein-glutamine gamma-glutamyltransferase 5; PSS2; TG(X); TGase 5; TGase X; TGASE5; TGase-5; TGASEX; Tgm5; TGM6; TGMX; TGX; transglutaminase 5; transglutaminase V; transglutaminase X; Transglutaminase-5 | |
Rabbit | |
Affinity chromatography | |
RUO | |
9333 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
O43548 | |
Tgm5 | |
Synthetic peptide directed towards the C-terminal of human TGM5 (aa 673-720). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction