Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TGM5 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA596558
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA596558 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA596558 Supplier Invitrogen™ Supplier No. PA596558
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: LQKLKARSFH GSQRGAELQP SRPTSLSQDS PRSLHTPSLR PSDVVQVSLK FKLLDPPNMG QDICFVLLAL NMSSQFKDLK VNLSAQSLLH DGSPLSPFWQ DTAFITLSPK EAKTYPCKIS YSQYSQYLST DKLIRISALG EEKSSPEKIL VNKIITLSYP SITINVLGAA VVNQPLSIQV IFSNPLSEQV EDCVLTVEGS GLFKKQQKVF LGVLKPQHQA SIILETVPFK SGQRQIQANM RSNKFKDIKG YRNVYVDFAL; Positive Samples: Mouse liver, Mouse heart, Rat liver, Rat heart; Cellular Location: Cytoplasm.

This gene encodes a member of the transglutaminase family. The encoded protein catalyzes formation of protein cross-links between glutamine and lysine residues, often resulting in stabilization of protein assemblies. This reaction is calcium dependent. Mutations in this gene have been associated with acral peeling skin syndrome.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TGM5
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.4 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene Tgm5
Gene Accession No. O43548, Q9D7I9
Gene Alias 2310007C07Rik; MGC141907; protein-glutamine gamma-glutamyltransferase 5; PSS2; TG(X); TGase 5; TGase X; TGASE5; TGase-5; TGASEX; Tgm5; TGM6; TGMX; TGX; transglutaminase 5; transglutaminase V; transglutaminase X; Transglutaminase-5
Gene Symbols Tgm5
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 461-720 of human TGM5 (NP_9639252).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 691929, 74176, 9333
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.